4-Brom-2-fluorthioanisol 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD251578-1GEA 61 EUR
BLDPBD251578-1G BLDPBD251578-10G BLDPBD251578-5G
4-Brom-2-fluorthioanisol 98%
4-Brom-2-fluorthioanisol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₆BrFS
Molecular Weight: 221,0899 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12756455
CAS-Nummer: 221030-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR