5-Brom-8-(trifluormethoxy)chinolin 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD251505-1GEA 224 EUR
BLDPBD251505-1G BLDPBD251505-100MG BLDPBD251505-250MG
5-Brom-8-(trifluormethoxy)chinolin 98%
5-Bromo-8-(trifluoromethoxy)quinoline

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₅BrF₃NO
Molecular Weight: 292,0520 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11504881
CAS-Nummer: 1065074-23-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR