5-Methoxypyrimidine-2-carbonitrile 95%

BLDPBD251279-100MGEA 61 EUR
BLDPBD251279-100MG
5-Methoxypyrimidine-2-carbonitrile 95%
5-Methoxypyrimidine-2-carbonitrile

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₅N₃O
Molecular Weight: 135,123 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00223675
CAS-Nummer: 87362-32-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR