Methyl 5-Chlorothiophene-2-carboxylate 98%

BLDPBD250374-100GEA 277 EUR
BLDPBD250374-100G BLDPBD250374-5G BLDPBD250374-1G BLDPBD250374-25G
Methyl 5-Chlorothiophene-2-carboxylate 98%
Methyl 5-Chlorothiophene-2-carboxylate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₆H₅ClO₂S
Molecular Weight: 176,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06203698
CAS-Nummer: 35475-03-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR