5-(4-Fluorphenyl)-3-pyridincarbaldehyd 95%

BLDPBD250369-1GEA 122 EUR
BLDPBD250369-1G BLDPBD250369-5G
5-(4-Fluorphenyl)-3-pyridincarbaldehyd 95%
5-(4-Fluorphenyl)-3-pyridincarbaldehyd

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₂H₈FNO
Molecular Weight: 201,2000 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD02093924
CAS-Nummer: 381684-96-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR