5H-Pyrido[4,3-b]indole 98%

BLDPBD248929-5GEA 286 EUR
BLDPBD248929-5G BLDPBD248929-0.025 BLDPBD248929-250MG BLDPBD248929-1G BLDPBD248929-100MG BLDPBD248929-25G
5H-Pyrido[4,3-b]indole 98%
5H-Pyrido[4,3-b]indole

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₈N₂
Molecular Weight: 168,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00965976
CAS-Nummer: 244-69-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR