Ethylimidazo[1,2-a]pyridin-7-carboxylat 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD248213-25GEA 359 EUR
BLDPBD248213-25G BLDPBD248213-1G BLDPBD248213-5G
Ethylimidazo[1,2-a]pyridin-7-carboxylat 98%
Ethylimidazo[1,2-a]pyridin-7-carboxylat

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190,1986 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02325394
CAS-Nummer: 372147-49-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR