(2R,3R)-3-((S)-1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)-3-methoxy-2-methylpropanoic acid 95%

BLDPBD247924-250MGEA 125 EUR
BLDPBD247924-250MG BLDPBD247924-100MG
(2R,3R)-3-((S)-1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)-3-methoxy-2-methylpropanoic acid 95%
(2R,3R)-3-((S)-1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)-3-methoxy-2-methylpropanoic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₄H₂₅NO₅
Molecular Weight: 287,352 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18157735
CAS-Nummer: 120205-50-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR