2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-didecylfluorene 98%

BLDPBD247423-100MGEA 59 EUR
BLDPBD247423-100MG BLDPBD247423-1G BLDPBD247423-250MG
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-didecylfluorene 98%
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-didecylfluorene

Storage condition: Sealed in dry, Room Temperature

Formel: C₄₅H₇₂B₂O₄
Molecular Weight: 698,69 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20275090
CAS-Nummer: 711026-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR