9-n-Octyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole 99%

BLDPBD247406-1GEA 119 EUR
BLDPBD247406-1G
9-n-Octyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole 99%
9-n-Octyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₃₂H₄₇B₂NO₄
Molecular Weight: 531,35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23703113
CAS-Nummer: 478706-06-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR