N,N,N',N'-Tetrakis[4-(dibutylamino)phenyl]-1,4-phenylenediamine 98%

BLDPBD246842-5GEA 201 EUR
BLDPBD246842-5G BLDPBD246842-1G BLDPBD246842-250MG
N,N,N',N'-Tetrakis[4-(dibutylamino)phenyl]-1,4-phenylenediamine 98%
N,N,N',N'-Tetrakis[4-(dibutylamino)phenyl]-1,4-phenylenediamine

Storage condition: Sealed in dry, Room Temperature

Formel: C₆₂H₉₂N₆
Molecular Weight: 921,46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832942
CAS-Nummer: 4182-80-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR