3-Brom-7-chlorimidazo[1,2-a]pyridin 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD246467-1GEA 141 EUR
BLDPBD246467-1G BLDPBD246467-250MG BLDPBD246467-25G BLDPBD246467-0.025 BLDPBD246467-5G BLDPBD246467-100MG
3-Brom-7-chlorimidazo[1,2-a]pyridin 98%
3-Brom-7-chlorimidazo[1,2-a]pyridin

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₄BrClN₂
Molecular Weight: 231,4800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11111834
CAS-Nummer: 342613-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR