3-Brom-5-iodbenzonitril 97%

BLDPBD245413-1GEA 223 EUR
BLDPBD245413-1G BLDPBD245413-250MG BLDPBD245413-5G BLDPBD245413-100MG
3-Brom-5-iodbenzonitril 97%
3-Brom-5-iodbenzonitril

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₃BrIN
Molecular Weight: 307,9139 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00672949
CAS-Nummer: 289039-21-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR