2-Amino-5-brom-4-methoxypyridin 95%

BLDPBD243945-1GEA 152 EUR
BLDPBD243945-1G BLDPBD243945-100MG BLDPBD243945-5G BLDPBD243945-250MG
2-Amino-5-brom-4-methoxypyridin 95%
2-Amino-5-bromo-4-methoxypyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₇BrN₂O
Molecular Weight: 203,0400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15143383
CAS-Nummer: 1232431-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR