D(-)-Glutamin 96%

Synonyme: D(-)-2-Amino-1,5-pentandisäure-5-amidD(-)-Glutaminsäure-5-amid(R)-(-)-Glutamin(R)-(-)-2-Amino-1,5-pentandisäure-5-amid(R)-(-)-Glutaminsäure-5-amidH-D-Gln-OH

BLDPBD2435-5GEA 25 EUR
BLDPBD2435-5G BLDPBD2435-10G BLDPBD2435-100G BLDPBD2435-25G
D(-)-Glutamin 96%
D(-)-Glutamin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₅H₁₀N₂O₃
Molecular Weight: 146,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00065607
CAS-Nummer: 5959-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR