2-(tert-Butyldimethylsilyloxy)-4-bromanisol 98%
|
|
BLDPBD242798-1GEA
45
EUR
BLDPBD242798-1G
BLDPBD242798-5G
2-(tert-Butyldimethylsilyloxy)-4-bromanisol 98%
2-(tert-Butyldimethylsilyloxy)-4-bromanisol
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₁₃H₂₁BrO₂Si Molecular Weight: 317,2941 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11855959 CAS-Nummer: 177329-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...