4-(2,2,2-Trifluorethylcarbamoyl)phenylboronsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD240142-250MGEA 48 EUR
BLDPBD240142-250MG BLDPBD240142-1G BLDPBD240142-100MG BLDPBD240142-5G
4-(2,2,2-Trifluorethylcarbamoyl)phenylboronsäure 98%
4-(2,2,2-Trifluorethylcarbamoyl)phenylboronsäure

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₉BF₃NO₃
Molecular Weight: 246,9800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08235087
CAS-Nummer: 874459-90-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR