1-Tosyl-1,2,3,6-tetrahydropyridine-4-boronic acid, pinacol ester 97%

BLDPBD240125-1GEA 66 EUR
BLDPBD240125-1G BLDPBD240125-5G
1-Tosyl-1,2,3,6-tetrahydropyridine-4-boronic acid, pinacol ester 97%
1-Tosyl-1,2,3,6-tetrahydropyridine-4-boronic acid, pinacol ester

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₁₈H₂₆BNO₄S
Molecular Weight: 363,279 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD17926518
CAS-Nummer: 1256360-46-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR