1-Methyl-1H-indazol-4-boronsäure Hydrochlorid 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD239875-250MGEA 66 EUR
BLDPBD239875-250MG BLDPBD239875-100MG
1-Methyl-1H-indazol-4-boronsäure Hydrochlorid 95%
1-Methyl-1H-indazol-4-boronsäure Hydrochlorid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₁₀BClN₂O₂
Molecular Weight: 212,4400 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13176537
CAS-Nummer: 1257527-53-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR