6-Amino-3-bromchinolin 98%

BLDPBD239486-250MGEA 63 EUR
BLDPBD239486-250MG BLDPBD239486-5G BLDPBD239486-1G BLDPBD239486-100MG
6-Amino-3-bromchinolin 98%
6-Amino-3-bromoquinoline

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₉H₇BrN₂
Molecular Weight: 223,07 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07757603
CAS-Nummer: 7101-96-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR