Methyl 5-bromo-2-formylbenzoate 95%
|
|
BLDPBD238234-250MGEA
41
EUR
BLDPBD238234-250MG
BLDPBD238234-5G
BLDPBD238234-100MG
BLDPBD238234-10G
BLDPBD238234-1G
Methyl 5-bromo-2-formylbenzoate 95%
Methyl 5-bromo-2-formylbenzoate
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₉H₇BrO₃ Molecular Weight: 243,054 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD16036575 CAS-Nummer: 1016163-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...