2-(4-Bromophenyl)-4,6-diphenyl-1,3,5-triazine 98%

BLDPBD238071-100GEA 267 EUR
BLDPBD238071-0.025 BLDPBD238071-100G BLDPBD238071-250MG BLDPBD238071-1G BLDPBD238071-10G BLDPBD238071-5G BLDPBD238071-25G
2-(4-Bromophenyl)-4,6-diphenyl-1,3,5-triazine 98%
2-(4-Bromophenyl)-4,6-diphenyl-1,3,5-triazine

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₁H₁₄BrN₃
Molecular Weight: 388,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00194632
CAS-Nummer: 23449-08-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR