5-Iodo-2-nitrophenol 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD237766-100MGEA 39 EUR
BLDPBD237766-100MG BLDPBD237766-250MG BLDPBD237766-1G BLDPBD237766-5G
5-Iodo-2-nitrophenol 98%
5-Iodo-2-nitrophenol

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₆H₄INO₃
Molecular Weight: 265,005 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12923199
CAS-Nummer: 27783-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR