2-Methyl-2H-indazol-6-boronsäure 95+%

Lieferant: BLD PHARMATECH GMBH

BLDPBD237377-250MGEA 221 EUR
BLDPBD237377-250MG BLDPBD237377-100MG
2-Methyl-2H-indazol-6-boronsäure 95+%
2-Methyl-2H-indazole-6-boronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₉BN₂O₂
Molecular Weight: 175,9800 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09870057
CAS-Nummer: 1001907-57-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR