2-([1,1'-Biphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

BLDPBD236455-1GEA 7 EUR
BLDPBD236455-1G BLDPBD236455-5G BLDPBD236455-0.025 BLDPBD236455-10G BLDPBD236455-250MG BLDPBD236455-25G
2-([1,1'-Biphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-([1,1'-Biphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₈H₂₁BO₂
Molecular Weight: 280,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18452169
CAS-Nummer: 912844-88-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR