5-Bromo-3-chloroisoquinoline 95%

BLDPBD236384-100MGEA 49 EUR
BLDPBD236384-100MG BLDPBD236384-1G BLDPBD236384-5G BLDPBD236384-250MG
5-Bromo-3-chloroisoquinoline 95%
5-Bromo-3-chloroisoquinoline

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₅BrClN
Molecular Weight: 242,5 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11044700
CAS-Nummer: 1029720-67-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR