Ethyl-6-methylimidazo[1,2-a]pyridin-2-carboxylat 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD236353-100MGEA 30 EUR
BLDPBD236353-100MG BLDPBD236353-1G BLDPBD236353-250MG BLDPBD236353-5G
Ethyl-6-methylimidazo[1,2-a]pyridin-2-carboxylat 98%
Ethyl-6-methylimidazo[1,2-a]pyridin-2-carboxylat

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₂N₂O₂
Molecular Weight: 204,2300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02325396
CAS-Nummer: 70705-30-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR