4-Fluor-3-methoxyphenylboronsäurepinakolester 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD236257-5GEA 254 EUR
BLDPBD236257-5G BLDPBD236257-250MG BLDPBD236257-1G
4-Fluor-3-methoxyphenylboronsäurepinakolester 98%
4-Fluor-3-methoxyphenylboronsäurepinakolester

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₃H₁₈BFO₃
Molecular Weight: 252,09 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11227055
CAS-Nummer: 425378-85-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR