2-Chlor-4-(trifluormethoxy)phenylboronsäure 98%
|
|
BLDPBD236254-250MGEA
132
EUR
BLDPBD236254-250MG
BLDPBD236254-5G
BLDPBD236254-100MG
BLDPBD236254-1G
2-Chlor-4-(trifluormethoxy)phenylboronsäure 98%
2-Chlor-4-(trifluormethoxy)phenylboronsäure
Storage condition: Sealed in dry, 2-8°C
Formel:
C₇H₅BClF₃O₃ Molecular Weight: 240,3720 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11040272 CAS-Nummer: 345226-20-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...