3-Isopropylphenylboronsäurepinakolester 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD236249-5GEA 248 EUR
BLDPBD236249-5G BLDPBD236249-250MG BLDPBD236249-1G
3-Isopropylphenylboronsäurepinakolester 98%
3-Isopropylphenylboronsäurepinakolester

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₅H₂₃BO₂
Molecular Weight: 246,1529 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13195750
CAS-Nummer: 325142-89-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR