2-Ethoxy-5-pyrimidinboronsäure 95%

BLDPBD23594-1GEA 43 EUR
BLDPBD23594-1G BLDPBD23594-5G BLDPBD23594-250MG BLDPBD23594-100MG BLDPBD23594-10G
2-Ethoxy-5-pyrimidinboronsäure 95%
2-Ethoxy-5-pyrimidineboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₉BN₂O₃
Molecular Weight: 167,9600 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07375134
CAS-Nummer: 1003043-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR