Biotin-pentafluorphenylester 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD235214-250MGEA 69 EUR
BLDPBD235214-250MG
Biotin-pentafluorphenylester 95%
Biotin-pentafluorphenylester

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₆H₁₅F₅N₂O₃S
Molecular Weight: 410,36 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD09952636
CAS-Nummer: 120550-35-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR