(2S,6S)-tert-Butyl 2,6-dimethylpiperazine-1-carboxylate 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD233798-5GEA 458 EUR
BLDPBD233798-5G BLDPBD233798-25G BLDPBD233798-1G BLDPBD233798-250MG
(2S,6S)-tert-Butyl 2,6-dimethylpiperazine-1-carboxylate 98%
(2S,6S)-tert-Butyl 2,6-dimethylpiperazine-1-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₁H₂₂N₂O₂
Molecular Weight: 214,305 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11973745
CAS-Nummer: 574007-66-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR