Levodropropizine 97%

BLDPBD23352-5GEA 21 EUR
BLDPBD23352-5G BLDPBD23352-100G BLDPBD23352-1G BLDPBD23352-25G
Levodropropizine 97%
Levodropropizine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₃H₂₀N₂O₂
Molecular Weight: 236,32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00866852
CAS-Nummer: 99291-25-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR