4-Fluor-N-methylbenzamid 98%
Lieferant: BLD PHARMATECH GMBH
|
|
BLDPBD232218-10GEA
105
EUR
BLDPBD232218-10G
BLDPBD232218-1G
BLDPBD232218-5G
BLDPBD232218-250MG
BLDPBD232218-25G
4-Fluor-N-methylbenzamid 98%
4-Fluor-N-methylbenzamid
Storage condition: Sealed in dry, 2-8°C
Formel:
C₈H₈FNO Molecular Weight: 153,1536 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00032531 CAS-Nummer: 701-49-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...