(R)-5-Bromo-3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]pyridin-2-amine 98%
|
|
BLDPBD232207-25GEA
508
EUR
BLDPBD232207-25G
BLDPBD232207-1G
BLDPBD232207-250MG
BLDPBD232207-5G
(R)-5-Bromo-3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]pyridin-2-amine 98%
(R)-5-Bromo-3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]pyridin-2-amine
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₁₃H₁₀BrCl₂FN₂O Molecular Weight: 380,04 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18207061 CAS-Nummer: 877399-00-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...