4-Hydroxy-3-methoxybenzeneboronic acid 95%

BLDPBD231595-1GEA 336 EUR
BLDPBD231595-1G BLDPBD231595-250MG BLDPBD231595-100MG BLDPBD231595-5G
4-Hydroxy-3-methoxybenzeneboronic acid 95%
4-Hydroxy-3-methoxybenzeneboronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₇H₉BO₄
Molecular Weight: 167,955 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD18397223
CAS-Nummer: 182344-21-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR