3-Fluor-4-(trifluormethyl)phenylboronsäurepinakolester 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD231542-5GEA 108 EUR
BLDPBD231542-5G BLDPBD231542-1G
3-Fluor-4-(trifluormethyl)phenylboronsäurepinakolester 98%
3-Fluor-4-(trifluormethyl)phenylboronsäurepinakolester

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₁₅BF₄O₂
Molecular Weight: 290,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878544
CAS-Nummer: 445303-67-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR