4-(Cyanmethoxy)phenylboronsäure 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD231472-1GEA 176 EUR
BLDPBD231472-1G BLDPBD231472-250MG BLDPBD231472-100MG
4-(Cyanmethoxy)phenylboronsäure 95%
4-(Cyanmethoxy)phenylboronsäure

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₈BNO₃
Molecular Weight: 176,9700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09908170
CAS-Nummer: 947533-23-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR