4-Chlor-3-hydroxyphenylboronsäure 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD231346-100MGEA 87 EUR
BLDPBD231346-100MG BLDPBD231346-5G BLDPBD231346-1G BLDPBD231346-250MG
4-Chlor-3-hydroxyphenylboronsäure 95%
4-Chlor-3-hydroxyphenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₆BClO₃
Molecular Weight: 172,3740 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD10697413
CAS-Nummer: 915201-06-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR