2-(Ethoxycarbonyl)furan-3-boronsäure 97%

BLDPBD230792-1GEA 341 EUR
BLDPBD230792-1G BLDPBD230792-100MG BLDPBD230792-250MG
2-(Ethoxycarbonyl)furan-3-boronsäure 97%
2-(Ethoxycarbonyl)furan-3-boronsäure

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₇H₉BO₅
Molecular Weight: 183,9544 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD12025973
CAS-Nummer: 1150114-62-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR