2-(1,3-Dioxolan-2-yl)ethylboronsäurepinakolester 98%
Lieferant: BLD PHARMATECH GMBH
|
|
BLDPBD230787-5GEA
617
EUR
BLDPBD230787-5G
BLDPBD230787-100MG
BLDPBD230787-1G
BLDPBD230787-250MG
2-(1,3-Dioxolan-2-yl)ethylboronsäurepinakolester 98%
2-(1,3-Dioxolan-2-yl)ethylboronsäurepinakolester
Storage condition: Inert atmosphere, Store in freezer, under -20°C
Formel:
C₁₁H₂₁BO₄ Molecular Weight: 228,10 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD03788722 CAS-Nummer: 1073354-07-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...