3-Chlor-5-(dimethylcarbamoyl)phenylboronsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD230770-250MGEA 66 EUR
BLDPBD230770-250MG BLDPBD230770-1G
3-Chlor-5-(dimethylcarbamoyl)phenylboronsäure 98%
3-Chlor-5-(dimethylcarbamoyl)phenylboronsäure
Formel: C₉H₁₁BClNO₃
Molecular Weight: 227,4500 g/mol
MDL: MFCD09800861
CAS-Nummer: 957120-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR