5-(Diethylcarbamoyl)-2-fluorphenylboronsäure 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD230692-5GEA 617 EUR
BLDPBD230692-5G BLDPBD230692-250MG BLDPBD230692-1G
5-(Diethylcarbamoyl)-2-fluorphenylboronsäure 97%
5-(Diethylcarbamoyl)-2-fluorphenylboronsäure

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₁H₁₅BFNO₃
Molecular Weight: 239,0500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08436045
CAS-Nummer: 874289-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR