N-Cyclopropyl-4-fluorbenzolsulfonamid 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD230444-5GEA 263 EUR
BLDPBD230444-5G BLDPBD230444-1G BLDPBD230444-250MG
N-Cyclopropyl-4-fluorbenzolsulfonamid 98%
N-Cyclopropyl-4-fluorbenzolsulfonamid

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₁₀FNO₂S
Molecular Weight: 215,2446 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02136916
CAS-Nummer: 425654-95-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR