2'-Anilino-6'-(dibutylamino)-3'-methylfluoran 97%

BLDPBD23017-100GEA 267 EUR
BLDPBD23017-100G BLDPBD23017-1G BLDPBD23017-500G BLDPBD23017-25G BLDPBD23017-5G BLDPBD23017-10G BLDPBD23017-250MG
2'-Anilino-6'-(dibutylamino)-3'-methylfluoran 97%
2'-Anilino-6'-(dibutylamino)-3'-methylfluoran

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₃₅H₃₆N₂O₃
Molecular Weight: 532,68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00307579
CAS-Nummer: 89331-94-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR