1-Boc-5-cyan-3-indolboronsäurepinakolester 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD230088-100MGEA 38 EUR
BLDPBD230088-100MG BLDPBD230088-1G BLDPBD230088-250MG BLDPBD230088-5G
1-Boc-5-cyan-3-indolboronsäurepinakolester 98%
1-Boc-5-cyan-3-indolboronsäurepinakolester

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₂₀H₂₅BN₂O₄
Molecular Weight: 368,2345 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD15143604
CAS-Nummer: 1185427-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR