1-Brom-2,4-dichlor-5-nitrobenzol 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD229528-1GEA 38 EUR
BLDPBD229528-1G BLDPBD229528-25G BLDPBD229528-5G
1-Brom-2,4-dichlor-5-nitrobenzol 98%
1-Bromo-2,4-dichloro-5-nitrobenzene

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₂BrCl₂NO₂
Molecular Weight: 270,9000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16038032
CAS-Nummer: 65001-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR