5-Amino-4-cyan-1-(2,4-difluorphenyl)-3-methyl-1H-pyrazol 98%

BLDPBD229335-1GEA 215 EUR
BLDPBD229335-1G BLDPBD229335-250MG BLDPBD229335-100MG
5-Amino-4-cyan-1-(2,4-difluorphenyl)-3-methyl-1H-pyrazol 98%
5-Amino-4-cyano-1-(2,4-difluorophenyl)-3-methyl-1H-pyrazole

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₁H₈F₂N₄
Molecular Weight: 234,2048 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08459254
CAS-Nummer: 1020057-92-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR