5-tert-Butyl-3-iod-1H-pyrazol 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD229317-250MGEA 40 EUR
BLDPBD229317-250MG BLDPBD229317-100MG BLDPBD229317-5G BLDPBD229317-25G BLDPBD229317-1G
5-tert-Butyl-3-iod-1H-pyrazol 98%
5-tert-Butyl-3-iod-1H-pyrazol

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₇H₁₁IN₂
Molecular Weight: 250,0801 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13195724
CAS-Nummer: 1218791-05-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR